Skip to Content
Merck
All Photos(1)

Key Documents

SMB00005

Sigma-Aldrich

Austdiol

≥95% (LC/MS-ELSD)

Synonym(s):

Austadiol, Austdiol, Austidiol

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€378.00

€378.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 MG
€378.00

About This Item

Empirical Formula (Hill Notation):
C12H12O5
CAS Number:
Molecular Weight:
236.22
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

€378.00


Usually ships in 2 weeks.

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C=C2[C@@H]([C@](C(=O)C(=C2C=C1C)C=O)(O)C)O

InChI

1S/C12H12O5/c1-6-3-7-8(4-13)10(14)12(2,16)11(15)9(7)5-17-6/h3-5,11,15-16H,1-2H3/t11-,12-/m0/s1

InChI key

QVMUHZHZYCDMAI-RYUDHWBXSA-N

General description

Natural product derived from fungal source.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonardo Lo Presti et al.
Acta crystallographica. Section C, Crystal structure communications, 59(Pt 4), O199-O201 (2003-04-12)
The title compound, (7R,8S)-7,8-dihydroxy-3,7-dimethyl-6-oxo-7,8-dihydro-6H-isochromene-5-carbaldehyde, C(12)H(12)O(5), is a trans-vicinal diol. Of the two fused rings, which lie approximately in the same plane, the pyran ring is almost perfectly planar, while the cyclohexenone ring adopts a slightly distorted half-chair conformation. The crystal
Leonardo Lo Presti et al.
The journal of physical chemistry. B, 110(12), 6405-6414 (2006-03-24)
The total experimental electron density rho(r), its Laplacian inverted delta(2)rho(r), the molecular dipole moment, the electrostatic potential phi(r), and the intermolecular interaction energies have been obtained from an extensive set of single-crystal X-ray diffracted intensities, collected at T = 70(1)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service