Skip to Content
Merck
All Photos(1)

Key Documents

632570

Sigma-Aldrich

4-Phenylthiazole-2-thiol

technical grade, 90%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€103.00

About This Item

Empirical Formula (Hill Notation):
C9H7NS2
CAS Number:
Molecular Weight:
193.29
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€103.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Assay

90%

form

solid

mp

167-172 °C

functional group

phenyl

SMILES string

Sc1nc(cs1)-c2ccccc2

InChI

1S/C9H7NS2/c11-9-10-8(6-12-9)7-4-2-1-3-5-7/h1-6H,(H,10,11)

InChI key

CYCKHTAVNBPQDB-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service