Skip to Content
Merck
All Photos(1)

Key Documents

SCP0210

Sigma-Aldrich

Pheromonotropin Pseudalatia

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€183.00

€183.00


Usually ships in 2 weeks.


Select a Size

Change View
1 MG
€183.00

About This Item

Empirical Formula (Hill Notation):
C101H155N25O30
Molecular Weight:
2199.46
UNSPSC Code:
12352202
NACRES:
NA.32

€183.00


Usually ships in 2 weeks.

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥65%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Lys-Leu-Ser-Tyr-Asp-Asp-Lys-Val-Phe-Glu-Asn-Val-Glu-Phe-Thr-Pro-Arg-Leu-NH2

Application

Pheromonotropin Pseudalatia is a pheromonotropic neuropeptide structurally related to leucopyrokinin from the armyworm larvae. Pheromonotropins are use to study the pheromone systems of insects.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service