Skip to Content
Merck
All Photos(1)

Key Documents

200433

Sigma-Aldrich

Polybutadiene, phenyl terminated

average Mn ~1,800

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
605258
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

mol wt

average Mn ~1,800

Quality Level

composition

cis-1,4, 5%
trans-1,4, 10%
vinyl, 45%

density

0.93 g/mL at 25 °C (lit.)
0.93 g/mL at 25 °C

SMILES string

C=CC=C

InChI

1S/C4H6/c1-3-4-2/h3-4H,1-2H2

InChI key

KAKZBPTYRLMSJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

contains 60% unsaturation

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service