Skip to Content
Merck
All Photos(1)

Key Documents

SCP0194

Sigma-Aldrich

MMP 3 Substrate

≥95% (HPLC), lyophilized

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€261.00

€261.00


Usually ships in 2 weeks.


Select a Size

Change View
5 MG
€261.00

About This Item

Empirical Formula (Hill Notation):
C54H65N13O14S1
Molecular Weight:
1152.24
UNSPSC Code:
12352204
NACRES:
NA.32

€261.00


Usually ships in 2 weeks.

Product Name

MMP 3 Substrate,

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥80%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

DNP-Pro-Tyr-Ala-Tyr-Trp-Met-Arg

Application

Matrix metalloproteinases (MMP) are a family of zinc-dependent endopeptidases that degrade matrix proteins. MMPs include collagenases, gelatinases, matrilysins, enamelysins, metalloelastases, stromelysins and other structural protein and receptor lysins. The activity of these enzymes is selectively inhibited or measured by appropriately labeled peptides. DNP-Pro-Tyr-Ala-Tyr-Trp-Met-Arg (DNP-PYAYWMR) is used as a fluorogenic substrate for matrix metalloprotein-3 (MMP-3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service