Skip to Content
Merck
All Photos(1)

Key Documents

SAE0093

Sigma-Aldrich

Beta-1,4-galactosyltransferase 1

B4GALT1 human recombinant, expressed in HEK 293 cells, 2000 units/mg protein

Synonym(s):

Beta-1,4-GalTase 1, Beta4Gal-T1, UDP-Gal:beta-GlcNAc beta-1,4-galactosyltransferase 1, UDP-galactose:beta-N-acetylglucosamine beta-1,4-galactosyltransferase 1, b4Gal-T1

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€472.00

€472.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
€472.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

€472.00


Check Cart for Availability

Request a Bulk Order

recombinant

expressed in HEK 293 cells

Assay

95% (SDS-PAGE)

form

lyophilized powder

specific activity

2000 units/mg protein

shipped in

ambient

storage temp.

−20°C

General description

Recombinant human Beta-1,4-galactosyltransferase 1 (B4GALT1) is expressed in human HEK 293 cells as a glycoprotein with a calculated molecular mass of 40 kDa. The DTT-reduced protein migrates as a ~55 kDa polypeptide on SDS-PAGE due to glycosylation. This protein is manufactured in human cells, with no serum. The human cells expression system allows human-like glycosylation and folding, and often supports higher specific activity of the protein.

Application

This recombinant B4GALT1 product can be used to study the mode of action of the enzyme, as well as its potential inhibitors. It can also be used as a glycoengineering tool to modify glycoproteins in vitro.

Biochem/physiol Actions

β(1→4) galactosyltransferase 1 (B4GALT1) is a type II membrane-bound glycoprotein that transfers galactose from uridine diphosphate-α-D-galactose (UDP-galactose) to acceptor sugars, such as N-Acetylglucosamine (GlcNAc), in a β(1→4) linkage. B4GALT1 resides in the Golgi apparatus of higher eukaryotic cells.
A major function of B4GALT1 is the addition of β(1→4) linked galactose residues to oligosaccharide acceptors with terminal N-acetylglucosamine residues. This is a late elongation step in the N-glycan processing pathway.B4GALT1 enzymatic activity is widely distributed in the vertebrate kingdom, in both mammals and non-mammals, including avians and amphibians.B4GALT1 enzymatic activity has also been demonstrated in a subset of plants which diverged from animals an estimated 1 billion years ago.B4GALT1 interacts with α-lactalbumin (LA), a protein expressed in the mammary gland during lactation, to form the lactose synthase (LS) complex that transfers galactose from UDP-α-D-Gal to glucose, producing the lactose secreted in milk.Defects in B4GALT1 are the cause of congenital disorder of glycosylation type 2D (CDG2D).Glomerular B4GALT1 expression has been found to be increased in IgA nephropathy. IgA binding and IgA-induced mesangial cell phosphorylation of spleen tyrosine kinase and IL-6 synthesis were inhibited by a panel of β(1→4) galactosyltransferase-specific antibodies, which suggests that IgA binds to the catalytic domain of β(1→4) galactosyltransferase.

Unit Definition

One unit is defined as the amount of enzyme required to transfer 1.0 nanomole of galactose from UDP-Gal to glucosamine per minute at pH 7.9, 37 oC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Verena Janes et al.
Cells, 9(3) (2020-03-07)
Genetic defects of human galactose-1-phosphate uridyltransferase (hGALT) and the partial loss of enzyme function result in an altered galactose metabolism with serious long-term developmental impairment of organs in classic galactosemia patients. In search for cellular pathomechanisms induced by the stressor

Articles

Explore tools for glycosyltransferase synthesis and modification of glycans, such as glycosyltransferases and nucleotide sugar donors.

Enzymatic glycosyltransferase specificity challenges the one enzyme-one linkage concept.

Questions

  1. この酵素のKm値を教えていただきたいです

    1 answer
    1. A Km value for this product is not available.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service