Skip to Content
Merck
All Photos(1)

Key Documents

233264

Millipore

CMP-Sialic Acid, Disodium Salt

CMP-sialic acid is the activated form of sialic acid that is required for the biosynthesis of sialic acid-containing complex carbohydrates.

Synonym(s):

CMP-Sialic Acid, Disodium Salt, CMP-N-Acetylneuraminic Acid, 2Na, CMP-NANA, CMP-Neu5Ac

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
Inquire
25 MG
Inquire


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
Inquire
25 MG
Inquire

About This Item

Empirical Formula (Hill Notation):
C20H29N4O16P · 2Na
CAS Number:
Molecular Weight:
658.41
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.79


Estimated to ship on01 June 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

water: 20 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

[P](=O)(OC3(OC(C(C(C3)O)NC(=O)C)C(O)C(O)CO)C(=O)O)(OCC1OC(C(C1O)O)N2C=CC(=NC2=O)N)O

InChI

1S/C20H31N4O16P/c1-7(26)22-12-8(27)4-20(18(32)33,39-16(12)13(29)9(28)5-25)40-41(35,36)37-6-10-14(30)15(31)17(38-10)24-3-2-11(21)23-19(24)34/h2-3,8-10,12-17,25,27-31H,4-6H2,1H3,(H,22,26)(H,32,33)(H,35,36)(H2,21,23,34)

InChI key

TXCIAUNLDRJGJZ-UHFFFAOYSA-N

General description

CMP-sialic acid is the activated form of sialic acid that is required for the biosynthesis of sialic acid-containing complex carbohydrates. It is a widespread carbohydrate with numerous biological functions, including blood protein half-life regulation, variety of toxin neutralization, cellular adhesion and glycoprotein lytic protection. Also available as a 25 mM solution in H2O (Cat. No. 505223).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Unstable under acidic conditions.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Enzymatic glycosyltransferase specificity challenges the one enzyme-one linkage concept.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service