Skip to Content
Merck
All Photos(1)

Key Documents

SAB5701767

Sigma-Aldrich

Anti-Phospho-α-Synuclein-S129 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€328.00

€328.00


Check Cart for Availability


Select a Size

Change View
100 μL
€328.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€328.00


Check Cart for Availability

biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human

concentration

1.35 mg/mL

technique(s)

western blot: 1:500-1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer129 )

Gene Information

human ... SNCA(6622)

General description

Alpha-synuclein is a member of the synuclein family, which also includes beta- and gamma-synuclein. Synucleins are abundantly expressed in the brain and alpha- and beta-synuclein inhibit phospholipase D2 selectively. SNCA may serve to integrate presynaptic signaling and membrane trafficking. Defects in SNCA have been implicated in the pathogenesis of Parkinson disease. SNCA peptides are a major component of amyloid plaques in the brains of patients with Alzheimer′s disease. Alternatively spliced transcripts encoding different isoforms have been identified for this gene.

Immunogen

A phospho specific peptide corresponding to residues surrounding S129 of human alpha-Synuclein

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service