Skip to Content
Merck
All Photos(1)

Key Documents

AB9850

Sigma-Aldrich

Anti-phospho-α Synuclein (Ser129) Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€544.00


Estimated to ship on30 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer129)

Gene Information

human ... SNCA(6622)

Specificity

Alpha Synuclein, phospho Serine 129. The antibody recognizes a protein of ~15 kDa corresponding to alpha synuclein, phospho Serine 129 in lysates from rat cortex. Immunolableing is blocked by preadsorption with the phospho-peptide used as the immunogen but not by the corresponding dephospho-peptide.
The immunogen has 100% homology with human, mouse, non-human primate, bovine and canine.

Immunogen

Synthetic peptide of amino acids surrounding the phosphoSerine 129 site of rat alpha synuclein.

Application

This Anti-phospho-α Synuclein (Ser129) Antibody is validated for use in WB for the detection of phospho-α Synuclein (Ser129).

Linkage

Replaces: 04-1052

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ningshan Wang et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 68(10), 669-678 (2020-09-15)
The detection of cutaneous phosphorylated alpha-synuclein (P-syn) in patients with Parkinson's disease (PD) has ranged from 30% to 100% across different studies. We hypothesize that part of the variability in P-syn detection is due to methodological differences using sections of
J L Barr et al.
British journal of pharmacology, 172(22), 5414-5424 (2015-09-17)
Ceftriaxone is a β-lactam antibiotic and glutamate transporter activator that reduces the reinforcing effects of psychostimulants. Ceftriaxone also reduces locomotor activation following acute psychostimulant exposure, suggesting that alterations in dopamine transmission in the nucleus accumbens contribute to its mechanism of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service