SAB5600176
Anti-Human Kappa Light Chain-Biotin antibody, Rabbit monoclonal
recombinant, expressed in HEK 293 cells, clone RM126, purified immunoglobulin
Synonym(s):
Anti-D4Mit8
About This Item
Recommended Products
recombinant
expressed in HEK 293 cells
conjugate
biotin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
RM126, monoclonal
recombinant monoclonal
form
buffered aqueous glycerol solution
species reactivity
human
concentration
1 mg/mL
technique(s)
ELISA: 0.05-0.2 μg/mL
immunofluorescence: 0.5-2 μg/mL
immunohistochemistry: 0.5-2 μg/mL
isotype
IgG
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service