Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700690

Sigma-Aldrich

Monoclonal Anti-CD209-PE antibody produced in mouse

clone UW60.1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CDSIGN, CLEC4L, DC-SIGN1, Anti-DC-SIGN

Sign Into View Organizational & Contract Pricing

Select a Size

100 TEST
€360.00

€360.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 TEST
€360.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

€360.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

UW60.1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD209(30835)

General description

The mouse monoclonal antibody UW60.1 recognizes human CD209 (DC-SIGN), a 44 kDa transmembrane receptor expressed on the surface of dendritic cells and macrophages.

Immunogen

CD209-His-tagged fusion protein

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shilei Liu et al.
Experimental and therapeutic medicine, 22(2), 871-871 (2021-07-02)
Atherosclerosis (AS) is a chronic pathophysiological process that causes high mortality and morbidity. It has previously been reported that homeobox A9 (HOXA9) may participate in regulation of the cardiovascular system and the pathology of AS by upregulating E-selectin and vascular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service