Skip to Content
Merck
All Photos(4)

Documents

SAB4501490

Sigma-Aldrich

Anti-GPR120 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G-protein coupled receptor 120, G-protein coupled receptor 129, G-protein coupled receptor GT01, G-protein coupled receptor PGR4, GPR120

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 42 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR120(338557)

General description

Anti-GPR120 Antibody detects endogenous levels of total GPR120 protein.
GPR120 (G-protein coupled receptor 120) is also called as free fatty acid receptor 4 (FFA4). It is present in mature bone cells. It is located on human chromosome 12q24.

Immunogen

The antiserum was produced against synthesized peptide derived from human GPR120.

Immunogen Range: 221-270

Application

Anti-GPR120 antibody has been used in immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

GPR120 (G-protein coupled receptor 120) promotes bone formation and suppresses bone resorption, thereby it plays an important role in bone metabolism.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Free Fatty Acid Receptor 4 (GPR120) Stimulates Bone Formation and Suppresses Bone Resorption in the Presence of Elevated n-3 Fatty Acid Levels.
Ahn SH, et al.
Endocrinology, 157(7), 2621-2635 (2016)
Patricia Widmayer et al.
Frontiers in physiology, 6, 53-53 (2015-03-17)
Long-term intake of dietary fat is supposed to be associated with adaptive reactions of the organism and it is assumptive that this is particularly true for fat responsive epithelial cells in the mucosa of the gastrointestinal tract. Recent studies suggest
Chromosome 12q24.31-q24.33 deletion causes multiple dysmorphic features and developmental delay: First mosaic patient and overview of the phenotype related to 12q24qter defects.
Al-Zahrani J, et al.
Molecular Cytogenetics, 4, 9-9 (2011)
Seong Hee Ahn et al.
Endocrinology, 157(7), 2621-2635 (2016-05-05)
Free fatty acid receptor 4 (FFA4) has been reported to be a receptor for n-3 fatty acids (FAs). Although n-3 FAs are beneficial for bone health, a role of FFA4 in bone metabolism has been rarely investigated. We noted that
Yufeng Zhao et al.
Molecular medicine reports, 17(2), 3281-3287 (2017-12-20)
Grifolic acid is a phenolic compound that was first extracted from the mushroom Albatrellus confluens; it acts as an agonist of the free fatty acid receptor (FFAR4). FFAR4 is expressed in macrophages and mediates the anti‑inflammatory effects of n‑3 unsaturated free

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service