Skip to Content
Merck
All Photos(2)

Key Documents

SAB4600179

Sigma-Aldrich

Anti-Chicken IgY (H+L), highly cross-adsorbed, CF 647 antibody produced in goat

~2 mg/mL, affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

50 μL
€104.00
250 μL
€203.00

€104.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
50 μL
€104.00
250 μL
€203.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€104.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

goat

Quality Level

conjugate

CF 647 conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

chicken

concentration

~2 mg/mL

technique(s)

flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect immunofluorescence: 1-10 μg/mL

fluorescence

λex 650 nm; λem 665 nm

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Immunoglobulin Y is the antibody specifically produced in birds. It is mostly related to mammalian IgG. However, IgY has different characteristics compared to mammalian IgG. For example, IgY does not bind to mammalian complements nor rheumatoid factor, which are present in mammalian serum and anti-mammalian IgG antibodies, which often cross-react to IgG from different mammalian species, exhibit low cross-reactivity to IgY. These characteristics of IgY is beneficial when it is used as a biochemical/clinical reagent. Immunoglobulin Y present in chicken blood serum has the capability to pass across the follicular epithelium of the ovary and aggregate in the yolk during oogenesis. Structurally, IgY and mammalian IgG differ in the heavy chain with the molecular weight of 68,000 and 50,000 respectively. Mammalian IgG has slightly higher stability compared to IgY.

Specificity

Binds all chicken IgYs (minimal cross-reaction with bovine, goat, guinea pig, syrian hamster, horse, human, mouse, rabbit, rat and sheep proteins)

Immunogen

chicken IgY (H+L)

Application

Anti-Chicken IgY (H+L), highly cross-adsorbed, CF 647 antibody produced in goat has been used in immunocytochemistry.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 2 mg/mL bovine serum albumin.

Preparation Note

Protect from light.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Default Patterning Produces Pan-cortical Glutamatergic and CGE/LGE-like GABAergic Neurons from Human Pluripotent Stem Cells
Floruta CM
Stem Cell Reports, 9, 1463-1476 (2017)
Molecular stability of chicken and rabbit immunoglobulin G
Shimizu M
Bioscience, Biotechnology, and Biochemistry, 56, 270-274 (1992)
Immunoglobulins of the non-galliform birds: antibody expression and repertoire in the duck.
Lundqvist ML
Developmental and Comparative Immunology, 30, 93-100 (2006)
Máté Fischer et al.
Alcoholism, clinical and experimental research, 45(5), 979-995 (2021-03-09)
Fetal alcohol syndrome (FAS) due to gestational alcohol exposure represents one of the most common causes of nonheritable lifelong disability worldwide. In vitro and in vivo models have successfully recapitulated multiple facets of the disorder, including morphological and behavioral deficits
Crina M Floruta et al.
Stem cell reports, 9(5), 1463-1476 (2017-11-07)
Default differentiation of human pluripotent stem cells has been promoted as a model of cortical development. In this study, a developmental transcriptome analysis of default-differentiated hPSNs revealed a gene expression program resembling in vivo CGE/LGE subpallial domains and GABAergic signaling. A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service