Skip to Content
Merck

Skip To

AC146

Normal Chicken IgY Antibody, purified | AC146

1 mg/mL, Chemicon®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 116,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Technique(s):
blocking: suitable (interfering antibodies)
Application:
blocking
Citations:
4

€ 116,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken

Quality Segment

antibody form

purified antibody

clone

polyclonal

manufacturer/tradename

Chemicon®

concentration

1 mg/mL

technique(s)

blocking: suitable (interfering antibodies)

isotype

IgY

shipped in

wet ice

target post-translational modification

unmodified

General description

Chickens transfer high quantities of IgY (IgG) into the egg yolk and harvesting antibodies from eggs eliminates the need for the invasive bleeding procedure. One week′s eggs can contain 10 times more antibodies than the volume of rabbit blood obtained from one weekly bleeding. However, there are some disadvantages when using certain chicken derived antibodies in immunoassays. Chicken IgY does not fix mammalian complement component C1 and it does not perform as a precipitating antibody using standard solutions.
~180 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Application

Gel Electrophoresis Analysis: 20 µg of reduced Chicken IgY from a representative lot was resolved by SDS-PAGE and visualized using total protein stain.

Note: To be used as a negative control for ChIP Ab+ MeCP2, Cat. No. 17-10491.

Note: For use as a negative control, perform experiments in parallel using primary antibody or normal serum at the same concentration. A signal obtained with the normal serum indicates a nonspecific interaction; any additional signal with the primary antibody indicates a specific antibody-antigen interaction.
Research Category
Secondary & Control Antibodies
Research Sub Category
Isotype Control Antibodies

Physical form

Format: Purified
Purified chicken polyclonal in buffer containing PBS with 0.05% sodium azide.
T-gel

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting using non-reduced Chicken IgY.

Western Blotting Analysis: 0.01 µg of this non-reduced Chicken IgY was detected by a Rabbit anti-Chicken secondary antibody.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A9046C2288C6409
clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

biological source

chicken

biological source

rabbit

biological source

rabbit

biological source

rabbit

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

technique(s)

blocking: suitable (interfering antibodies)

technique(s)

direct ELISA: 1:30,000, dot blot: 1:160,000 (indirect chemiluminescence), immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:1,000

technique(s)

indirect ELISA: 1:25,000

technique(s)

indirect ELISA: 1:150,000-1:250,000, quantitative precipitin assay: 2.5-5 mg/mL

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

isotype

IgY

isotype

-

isotype

-

isotype

-


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters