Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503508

Sigma-Aldrich

Anti-WAVE1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KIAA0269, SCAR1, Verprolin homology domain-containing protein 1, WAS1, WASF1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Check Cart for Availability


Select a Size

Change View
100 μG
€463.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WASF1(8936)

General description

Wiskott–aldrich syndrome protein family member 1 (WAVE1) is encoded by the gene mapped to human chromosome 6q21-22. It is primarily expressed in brain and is characterized with a tripartite VCA (verprolin homology, cofilin homology and acidic) domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human WAVE1.

Immunogen Range: 91-140

Application

Anti-WAVE1 antibody produced in rabbit has been used in immunoblotting.[1]

Biochem/physiol Actions

Members of WASP-family verprolin-homologous protein (WAVE) family plays a vital role in actin polymerisation and cytoskeletal organisation, which are essential for various cellular processes, including cell shape changes, cytokinesis, cell motility and membrane traffic. WAVE1 regulates chemoresistance during autophagy. The encoded protein plays an essential role in oligodendrocyte morphogenesis and myelination. In addition, it is also implicated in regulation of spine morphogenesis. Overexpression of the gene leads to the development of prostate cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Knockdown of WAVE1 enhances apoptosis of leukemia cells by downregulating autophagy.
Zhang Z, et al.
International Journal of Oncology, 48(6), 2647-2656 (2016)
Phosphorylation of WAVE1 regulates actin polymerization and dendritic spine morphology.
Kim Y, et al.
Nature, 442(7104), 814-814 (2006)
Identification of two human WAVE/SCAR homologues as general actin regulatory molecules which associate with the Arp2/3 complex.
Suetsugu S, et al.
Biochemical and Biophysical Research Communications, 260(1), 296-302 (1999)
Expression of the WASP verprolin-homologues (WAVE members) in human breast cancer.
Fernando H S, et al.
Oncology, 73(5-6), 376-383 (2007)
WAVE1 is required for oligodendrocyte morphogenesis and normal CNS myelination.
Kim H J, et al.
The Journal of Neuroscience, 26(21), 5849-5859 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service