Skip to Content
Merck
All Photos(1)

Documents

SAB4502835

Sigma-Aldrich

Anti-SOX12 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

sox12, sox12 protein, sox22, sox22 protein, sry (sex determining region y)-box 12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 34 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX12(6666)

General description

Anti-SOX12 Antibody detects endogenous levels of total SOX12 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human SOX12.

Immunogen Range: 71-120

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Wang et al.
Oncology letters, 20(6), 389-389 (2020-11-17)
Colorectal cancer (CRC) is the third most common malignant type of tumor worldwide. Neurensin-2 (NRSN2) is a small neuronal membrane protein associated with tumorigenesis. Therefore, the present study aimed to investigate the association between NRSN2 and CRC, and further examined
Feng Du et al.
Cell death & disease, 10(3), 239-239 (2019-03-13)
The sex-determining region Y (SRY)-box (SOX) family has a crucial role in carcinogenesis and cancer progression. However, the role of SOX12 and the mechanism by which it is dysregulated in colorectal cancer (CRC) remain unclear. Here we analyzed SOX12 expression
Chunguang Li et al.
Oncology reports, 45(1), 129-138 (2021-01-09)
As a crucial transcription factor, sex‑determining region Y box 12 (SOX12) is closely related with tumorigenesis and malignant transformation in various malignant tumor types. To date, the specific function of SOX12 in esophageal squamous cell carcinoma (ESCC) has remained largely

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service