Merck
  • SOX12 promotes colorectal cancer cell proliferation and metastasis by regulating asparagine synthesis.
SOX12 promotes colorectal cancer cell proliferation and metastasis by regulating asparagine synthesis.
Cell death & disease (2019-03-13)
Feng Du, Jie Chen, Hao Liu, Yanhui Cai, Tianyu Cao, Weili Han, Xiaofang Yi, Meirui Qian, Dean Tian, Yongzhan Nie, Kaichun Wu, Daiming Fan, Limin Xia
ABSTRACT
The sex-determining region Y (SRY)-box (SOX) family has a crucial role in carcinogenesis and cancer progression. However, the role of SOX12 and the mechanism by which it is dysregulated in colorectal cancer (CRC) remain unclear. Here we analyzed SOX12 expression patterns in two independent CRC cohorts (cohort I, n = 390; cohort II, n = 363) and found that SOX12 was significantly upregulated in CRC, indicating a poor prognosis in CRC patients. Overexpression of SOX12 promoted CRC cell proliferation and metastasis, whereas downregulation of SOX12 hampered CRC aggressiveness. Mechanistically, SOX12 facilitated asparagine synthesis by transactivating glutaminase (GLS), glutamic oxaloacetic transaminase 2 (GOT2), and asparagine synthetase (ASNS). Downregulation of GLS, GOT2, and ASNS blocked SOX12-mediated CRC cell proliferation and metastasis, whereas ectopic expression of GLS, GOT2, and ASNS attenuated the SOX12 knockdown-induced suppression of CRC progression. In addition, serial deletion, site-directed mutagenesis, luciferase reporter, and chromatin immunoprecipitation (ChIP) assays indicated that hypoxia-inducible factor 1α (HIF-1α) directly binds to the SOX12 promoter and induces SOX12 expression. Administration of L-asparaginase decreased SOX12-mediated tumor growth and metastasis. In human CRC samples, SOX12 expression positively correlated with GLS, GOT2, ASNS, and HIF-1α expression. Based on these results, SOX12 may serve as a prognostic biomarker and L-asparaginase represents a potential novel therapeutic agent for CRC.
Sigma-Aldrich
Anti-SOX12 antibody produced in rabbit, affinity isolated antibody