Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502262

Sigma-Aldrich

Anti-PPAR-γ antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NR1C3, PPAR-γ-1, Peroxisome proliferator activated receptor γ

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Check Cart for Availability


Select a Size

Change View
100 μG
€463.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 57 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PPARG(5468)

General description

Peroxisome proliferator-activated receptor γ (PPARγ) is one of the subtypes of PPAR, and belongs to the nuclear hormone receptor superfamily. PPARγ is further sub classified into four isoforms and they have tissue specific expression. PPARγ is encoded by the gene mapped to human chromosome 3p25. PPARγ contains N- terminal AF (activation function)-1 transactivation domain, followed by a DNA-binding domain and a dimerization and ligand-binding domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human PPAR-gamma.

Immunogen Range: 78-127

Biochem/physiol Actions

Peroxisome proliferator-activated receptor γ (PPARγ) induces the expression of genes coding for proteins implicated in glucose and lipid metabolism. Natural PPAR-γ agonists found in foods functions as an anti-inflammatory molecule, thus, PPARγ is not only a target of the pharmaceutical industry, but also a novel target of the food industry. PPAR-γ regulates energy storage, adipocyte differentiation, and cellular inflammatory and ischemic responses. PPARγ is also involved in the production of prostanoids that regulate vascular function. PPARγ agonists, thiazolidinediones including rosiglitazone and pioglitazone, enhance insulin sensitivity by inhibiting the tumor necrosis factor α (TNF-α) activity in adipocytes. These PPAR-γ agonists are used in the treatment of type 2 diabetes. PPARγ functions as a potential therapeutic target in pulmonary hypertension.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The peroxisome proliferator-activated receptor: A family of nuclear receptors role in various diseases.
Tyagi S
Journal of Advanced Pharmaceutical Technology & Research, 2(4), 236-240 (2011)
Beneficial effects of PPAR-gamma ligands in ischemia-reperfusion injury, inflammation and shock.
Abdelrahman M
Cardiovascular Research, 65(4), 772-781 (2005)
PPARgamma as a potential therapeutic target in pulmonary hypertension.
Sutliff RL
Therapeutic Advances in Respiratory Disease, 4(3), 143-160 (2010)
T Fedchenko et al.
PPAR research, 2022, 7537210-7537210 (2022-06-07)
The master clock, which is located in the suprachiasmatic nucleus (SCN), harmonizes clock genes present in the liver to synchronize life rhythms and bioactivity with the surrounding environment. The reversed feeding disrupts the expression of clock genes in the liver.
Adrián Jordá et al.
International journal of molecular sciences, 23(19) (2022-10-15)
Ranolazine (Rn) is a drug used to treat persistent chronic coronary ischemia. It has also been shown to have therapeutic benefits on the central nervous system and an anti-diabetic effect by lowering blood glucose levels; however, no effects of Rn

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service