Skip to Content
Merck
All Photos(1)

Key Documents

MAB3872

Sigma-Aldrich

Anti-PPAR γ Antibody, isoform 1&2

ascites fluid, clone 1H4, Chemicon®

Synonym(s):

PPAR gamma

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€379.00

€379.00


Estimated to ship on28 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1428


Select a Size

Change View
100 μL
€379.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€379.00


Estimated to ship on28 May 2025

A recombinant, preservative-free antibody is available for your target. Try ZRB1428

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1H4, monoclonal

species reactivity

human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPARG(5468)

General description

Peroxisome proliferator-activated receptors (PPARs) are nuclear receptors involved in lipid transport and metabolism. As such, their roles in chronic diseases such as diabetes, obesity, atherosclerosis and cancer are heavily investigated. Transcriptional activity of PPARs is regulated by fatty acid binding. Three PPAR isotypes have been identified: a, b and g. PPARg stimulates lipolysis of circulating triglycerides and the subsequent uptake of fatty acids into adipose cells. PPARs can bind to DNA only as a heterodimer with the retinoid X receptor (RXR).

Specificity

Reacts with human PPAR gamma2. The antibody also reacts with PPAR gamma1 due to immunogen sequence homology. The immunogen shows no homology with PPAR alpha or PPAR beta.

Immunogen

Epitope: isoform 1&2
Synthetic peptide from amino acids 107-121 of human PPAR gamma2.

Application

Anti-PPAR γ Antibody, isoform 1&2 is a Mouse Monoclonal Antibody for detection of PPAR gamma also known as Peroxisome Proliferator Activated Receptor γ & has been validated in ELISA, ICC & WB.

Target description

67 kDa

Analysis Note

Control
THP-1 cell lysate, 3T3-L1 cell lysate or U-937 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Younho Han et al.
Scientific reports, 6, 35655-35655 (2016-10-19)
Osterix is a novel bone-related transcription factor involved in osteoblast differentiation, and bone maturation. Because a reciprocal relationship exists between adipocyte and osteoblast differentiation of bone marrow derived mesenchymal stem cells, we hypothesized that Osterix might have a role in
Qi Tang et al.
International journal of molecular medicine, 40(3), 713-720 (2017-07-22)
Adipose tissue engraftment has become a well-established therapy in plastic and reconstructive surgery used to restore age-related or injury-related soft tissue loss. However, the unpredictable absorption rates limit its further application. Some clinicians have noted that more optimal aesthetic results
Yiyan Qiu et al.
Molecular medicine reports, 24(2) (2021-06-11)
Exercise intervention has become one of the most effective methods to prevent and treat osteoporosis, which is a common age‑related disease and seriously affects the health and quality of life of the elderly. However, the molecular mechanism remains to be
Yifeng Jin et al.
Scientific reports, 6, 34661-34661 (2016-10-04)
Conformational change in helix 12 can alter ligand-induced PPARγ activity; based on this reason, isoquinolinoquinazolinones, structural homologs of berberine, were designed and synthesized as PPARγ antagonists. Computational docking and mutational study indicated that isoquinolinoquinazolinones form hydrogen bonds with the Cys285
Christine Swanson et al.
Neurological research, 36(7), 634-646 (2014-03-14)
To characterize the distribution of peroxisome proliferator-activated receptor-gamma (PPAR-gamma) in the substantia nigra of normal and 1-methyl-4-phenyl-1,2,3,6-tetrahydropyridine (MPTP)-treated hemiparkinsonian monkeys, in order to validate PPAR-gamma as a target for neuroprotection. Immunohistochemical analysis of PPAR-gamma expression was performed in the substantia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service