Skip to Content
Merck
All Photos(2)

Documents

SAB4500799

Sigma-Aldrich

Anti-AKT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AKT, AKT1 kinase, C-AKT, PKB, kinase Akt1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKT1(207)

General description

Anti-AKT Antibody detects endogenous levels of total AKT protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Akt.

Immunogen Range: 212-261

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li-Tao Liu et al.
Molecular medicine reports, 17(1), 488-494 (2017-11-09)
Isolariciresinol-9'-O-α-L-arabinofuranoside (MWS‑19) isolated from Pinus massoniana Lamb. Fresh pine needles is the major ingredient of the Songling Xuemaikang capsule therapy used for hypertension. The present study aimed to investigate the effects and underlying mechanisms of MWS‑19 on hydrogen peroxide (H2O2)‑induced apoptosis
Longxing Cao et al.
Annals of translational medicine, 4(23), 462-462 (2017-01-17)
Thyroid cancer is very common, but skull ectopic thyroid cancer has not been reported in 50 years of literatures in foreign countries. There are only four cases of the skull ectopic thyroid cancer reported in more than 30 years of
Guanxue Xu et al.
Bioscience reports, 40(5) (2020-05-13)
Matrine is a main active constituent of Chinese herb Sophora flavescens Ait (Kushen), which has shown various pharmacological effects, and has been reported to exhibit protective effects in heart failure. In the present study, the underlying mechanism of matrine was
Xueliang Qin et al.
3 Biotech, 10(4), 187-187 (2020-04-08)
This study explored the effect of methyl-indole on pancreatic cancer cell viability and investigated the mechanism involved. The viability of pancreatic cells showed a significant suppression on treatment with methyl-indole in dose-based manner. Treatment with 5 µM methyl-indole suppressed Capan-1 cell
Simona Daniele et al.
Journal of neuroinflammation, 12, 225-225 (2015-12-03)
In the central nervous system, glial cells provide metabolic and trophic support to neurons and respond to protracted stress and insults by up-regulating inflammatory processes. Reactive astrocytes and microglia are associated with the pathophysiology of neuronal injury, neurodegenerative diseases and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service