Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301414

Sigma-Aldrich

Anti-phospho-Akt (pSer129) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AKT, AKT1 kinase, RAC, RAC-PK-alpha, kinase Akt1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€409.00

€409.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€409.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€409.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

65 kDa

species reactivity

rat, human, mouse

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001014431.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer129)

Gene Information

human ... AKT1(207)

Specificity

The antibody detects endogenous level of Akt only when phosphorylated at serine 129.

Immunogen

Peptide sequence around phosphorylation site of serine129(D-N-S(p)-G-A) derived from Human Akt .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qing-Hai Qu et al.
OncoTargets and therapy, 13, 7949-7961 (2020-09-05)
Long non-coding RNAs (lncRNAs) have been reported to play important roles in tumor biology. In this study, we aimed to investigate the effects of T-box transcription factor 5 antisense RNA 1 (TBX5-AS1) on aggressive phenotypes of non-small cell lung cancer
Xiaohua Xu et al.
Medical science monitor : international medical journal of experimental and clinical research, 24, 8064-8073 (2018-11-12)
BACKGROUND The FOLR2 gene encodes folate receptor-beta (FR-beta), which is expressed by tumor-associated macrophages. The effects of FOLR2 gene expression in non-small cell lung cancer (NSCLC) remains unknown. This study aimed to investigate the effects of FOLR2 gene expression and
Wencai Sun et al.
Experimental and therapeutic medicine, 15(1), 1119-1128 (2018-02-13)
The present study aimed to investigate the effect and underlying mechanism of microRNA (miR)-4262 in the development of osteoarthritis (OA) in rats. Primary chondrocytes were separated from Sprague-Dawley rats and then treated with tumor necrosis factor-α (TNF-α). The level of
Changsheng Shi et al.
Cancer biology & therapy, 21(7), 604-614 (2020-04-25)
Long noncoding RNAs (lncRNAs) have been reported to play a significant role in the occurrence and progression of tumors. In different tumors, they can either act as an oncogene or tumor suppressor via modulating various target mRNAs. OIP5-AS1 belongs to
Chunya Lu et al.
Journal of cellular and molecular medicine, 22(4), 2478-2487 (2018-02-03)
Lung cancer is the leading cause of death in individuals with malignant disease. Non-small-cell lung cancer (NSCLC) is the most common type of lung cancer, and chemotherapy drugs such as cisplatin are the most widely used treatment for this disease.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service