Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300122

Sigma-Aldrich

Anti-phospho-TYK2 (pTyr1054) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JTK1 antibody produced in rabbit, Anti-tyrosine kinase 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€485.00

€485.00


Check Cart for Availability


Select a Size

Change View
100 μG
€485.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€485.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~140 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(H-E-YP-Y-R)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1054)

Gene Information

human ... TYK2(7297)

Immunogen

Peptide sequence around phosphorylation site of tyrosine 1054 (H-E-Y(p)-Y-R), according to the protein TYK2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

TYK2 encodes a member of the tyrosine kinase and, more specifically, the Janus kinases (JAKs) protein families. This protein associates with the cytoplasmic domain of type I and type II cytokine receptors and promulgate cytokine signals by phosphorylating receptor subunits. It is also component of both the type I and type III interferon signaling pathways. As such, it may play a role in anti-viral immunity. A mutation in this gene has been associated with hyperimmunoglobulin E syndrome (HIES) - a primary immunodeficiency characterized by elevated serum immunoglobulin E.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yue Zhu Zhang et al.
Biomedical and environmental sciences : BES, 32(6), 406-418 (2019-07-03)
Previous studies have indicated that the plasticizer di (2-ethylhexyl) phthalate (DEHP) affects lipid accumulation; however, its underlying mechanism remains unclear. We aim to clarify the effect of DEHP on lipid metabolism and the role of TYK2/STAT1 and autophagy. In total
Xiaoyan Li et al.
Life sciences, 217, 283-292 (2018-12-15)
PMS1 Homolog 2, Mismatch Repair System Component Pseudogene 2 (PMS2L2) has been reported as an up-regulated long non-coding RNA (lncRNA) in osteoarthritis (OA) tissues. The purpose of the present work is to explore whether the differently expressed PMS2L2 is associated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service