Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200780

Sigma-Aldrich

Anti-Human IgM (μ-chain specific) antibody, Mouse monoclonal

clone MB-11, purified from hybridoma cell culture

Synonym(s):

Anti-Human immunoglobulin M

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€458.00

€458.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€458.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€458.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MB-11, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1.0 mg/mL

technique(s)

ELISA: 0.1-0.2 μg/mL using 2.5 μg/mL Human IgM myeloma for coating
immunoblotting: suitable

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Human IgM (m-chain specific) antibody, Mouse monoclonal (mouse IgG2b isotype) is derived from the MB-11 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a mouse immunized with purified human IgM protein.
Immunoglobulin M (IgM) is the major class of surface immunoglobulins on lymphocyte membranes. IgM is suggested to act as the first line of defense during microbial infections. IgM has a pentameric structure, in which monomers are linking together via disulfide bonds. Surface IgM is expressed on immature and mature B cells, whereas IgM heavy chain is expressed intracellularly in pre-B cells. IgM serves as the antigen receptor of naive B cells and is involved in B cell maturation and complement activation.

Specificity

Anti-Human IgM (m-chain specific) antibody, Mouse monoclonal specifically recognizes m chain of Human IgM. The antibody shows no cross-reactivity with human IgG, Fab, Fc, k, l, and g chains.

Immunogen

Purified human IgM

Application

The antibody is recommended to use in various immunological techniques, including ELISA and Immunoblot.

Biochem/physiol Actions

Immunoglobulin M (IgM) acts as an antigen specific part of the B cell antigen receptor on the surface of B lymphocytes that are not stimulated, in its monomeric form. Polymeric IgM molecules also serve as important activators of the classical complement cascade. IgM is essential in agglutination and cytolytic reactions. IgM serves as the antigen receptor of naive B cells and is involved in B cell maturation and complement activation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Vidarsson et al.
Infection and immunity, 66(6), 2866-2870 (1998-05-29)
Streptococcus pneumoniae is a major respiratory pathogen of infants, children, and the elderly. Polysaccharide vaccines have been useful in adult populations but do not elicit protective immunity in infants and young children. To enhance their immunogenicity, vaccines of pneumococcal polysaccharides

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service