Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200698

Sigma-Aldrich

Anti-MR-Pro ADM antibody, Mouse monoclonal

clone 2E12E6, purified from hybridoma cell culture

Synonym(s):

Midregional proadrenomedullin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€573.00

€573.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€573.00

About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

€573.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2E12E6, monoclonal

form

buffered aqueous solution

mol wt

antigen 5 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.25-0.5 μg/mL using human recombinant peptide of MR-Pro ADM (amino acid 45-92).
indirect ELISA: 30-60 ng/mL using human recombinant peptide of MR-Pro ADM (amino acid 45-92) for coating.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADM (133)

General description

Monoclonal Anti-MR-Pro ADM (mid-regional pro adrenomedullin) (mouse IgG1 isotype) is derived from the hybridoma 2E12E6 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with human recombinant protein. Mid-regional pro-adrenomedullin (MR-proADM) is obtained as a by-product of post-translational processing of prepro adrenomedullin (Adm, AM) peptide. This gene is located on human chromosome 11p15.

Immunogen

human MR-Pro ADM recombinant protein.

Application

Anti-MR-pro ADM has been used in immunoblotting (~5 kDa) and indirect enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Mid-regional pro-adrenomedullin (MR-proADM) has been suggested as a biomarker for plasma concentrations of adrenomedullin as it is stoichiometrically generated and relatively stable in the plasma. Upregulated levels of MR-pro ADM were associated with an increased risk of mortality and morbidity in patients with heart failure, independent of natriuretic peptides. It can stimulate vascular endothelial barrier function.

Physical form

a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrenomedullin surges are linked to acute episodes of the systemic capillary leak syndrome (Clarkson disease)
Xie Z, et al.
Journal of Leukocyte Biology, 103(4), 749-759 (2018)
Mid-Regional Pro-Adrenomedullin (MR-proADM) as a Biomarker for Sepsis and Septic Shock: Narrative Review
Healthcare (Basel, Switzerland) , 6(3), 110-110 (2018)
Elevated levels of adrenomedullin in eutopic endometrium and plasma from women with endometriosis
Matson BC, et al.
Fertility and Sterility, 109(6), 1072-1078 (2018)
Mihael Potocki et al.
Current heart failure reports, 9(3), 244-251 (2012-06-27)
Circulating biomarkers have become increasingly important in diagnosing and risk-stratifying patients with heart failure (HF). While the natriuretic peptides have received much focus in the past decade, there is increasing interest in the role of other circulating biomarkers such as
Sex-specific effects of NLRP6/AVR and ADM loci on susceptibility to essential hypertension in a Sardinian population
Glorioso N, et al.
PLoS ONE, 8(10), e77562-e77562 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service