Skip to Content
Merck
All Photos(2)

Key Documents

ABT334

Sigma-Aldrich

Anti-TSLP receptor Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

TSLP Receptor Antibody, TSLP Receptor, Thymic stromal lymphopoietin receptor, CRL2, ILXR, TSLPR, Cytokine receptor-like factor 2, Cytokine receptor-like 2, IL-XR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€414.00

€414.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€414.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€414.00


Estimated to ship on28 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TSLP(85480)

General description

Thymic stromal lymphopoietin (TSLP) receptor is a member of the Type I cytokine receptor family (type 5 subfamily). TSLP has been shown to play an important role in dendritic cell maturation and activation. Additionally, TSLP receptor is involved in the positive selection of regulatory T cells, induction of CD4+ T cell-mediated allergic reaction and maintenance of peripheral CD4+ T cell homeostasis. Furthermore, TSLP receptor is implicated in the development of the hematopoietic system. TSLP receptor forms a functional complex with TSLP receptor and IL7R which is capable of stimulating cell proliferation through activation of STAT3 and STAT5. TSLP receptor is primarily expressed in dendrites and monocytes. Additionally, it is expressed in heart, skeletal muscle, kidney and adult and fetal liver; and weakly expressed in T-cells.

Immunogen

KLH-conjugated linear peptide corresponding to human TSLP receptor.

Application

Detect TLSP Receptor using this rabbit polyclonal antibody, Anti-TSLP receptor Antibody validated for use in western blotting, IHC (Paraffin) & Immunofluorescence.
Immunohistochemistry Analysis: 5 µg/mL from a representative lot detected TSLP receptor in human liver tissue.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected TSLP receptor in human liver cells.
Research Category
Signaling
Research Sub Category
Cytokines & Cytokine Receptors

Quality

Evaluated by Western Blotting in human liver tissue lysate.

Western Blotting Analysis: 2 µg/mL of this antibody detected TSLP receptor in 15 µg of human liver tissue lysate.

Target description

~53 kDa observed. Uncharacterized bands may appear in some lysate(s). Uniprot describes 2 isoforms at ~27 kDa and ~42 kDa

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human liver tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service