Skip to Content
Merck
All Photos(6)

Key Documents

SAB3501176

Sigma-Aldrich

Anti-CXCR4 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€370.00

€370.00


Check Cart for Availability


Select a Size

Change View
100 μG
€370.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€370.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCR4(7852)

Looking for similar products? Visit Product Comparison Guide

General description

CXCR4, a G-protein coupled receptor (GPCR) with seven transmembrane domains, is a CXC chemokine receptor specific for stromal-derived-factor-1 (SDF-1 or CXCL12). CXCR4 was initially discovered as one of the co-receptors for HIV entry into CD4+ T cells. Blocking CXCR4 could be potentially used as novel therapeutics for HIV treatment.CXCR4 signaling plays an important role in the migration, proliferation and quiescence of hematopoietic stem cell and their retention within the bone marrow, where it has high levels of SDF-1/CXCL12. It has been demonstrated that CXCR4 signaling mediates CD20 up-regulation on B cells.CXCR4 is highly expressed in more than 23 types of cancer, including breast cancer, ovarian cancer, melanoma, and prostate cancer, while there is very less or no expression of CXCR4 in healthy tissues. CXCR4 expression in cancer cells has been reported to be associated with tumor survival, growth and metastasis in tissues with high levels of SDF-1/CXCL12, such as lungs, liver and bone marrow (4,5).CXCR4 has been shown to regulate neuronal migration, cell positioning and axon wiring (6,7). CXCR4 mutant mice displayed aberrant neuronal distribution, which implicates the role in neuronal disorders such as epilepsy. CXCR4 is also involved in WHIM syndrome. WHIM mutations in CXCR4 were recently found in patients with Waldenstrom′s macroglobulinemia, and these mutations are correlated to clinical resistance to ibrutinib (9,10).

Immunogen

Anti-CXCR4 antibody was raised against a peptide corresponding to 15 amino acids near the center of human CXCR4 isoform b. The immunogen is located within amino acids 170-220 of CXCR4.

Application

Antibody validated: Western Blot in human, mouse, and rat samples; Immunohistochemistry and Immunofluorescence in human samples. All other applications and species not yet tested.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service