Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500585

Sigma-Aldrich

Anti-SYNPO antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Synaptopodin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€408.00

€408.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€408.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€408.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SYNPO(11346)

Immunogen

SYNPO antibody was raised against a 17 amino acid peptide near the carboxy terminus of human SYNPO.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500585.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinwang Zhu et al.
International journal of molecular medicine, 53(3) (2024-01-19)
The senolytics dasatinib and quercetin (DQ) alleviate age‑related disorders. However, limited information is available regarding the effects of DQ on diabetic kidney disease (DKD). The present study aimed to explore the effects of DQ on DKD and its potential molecular
Sandeep K Mallipattu et al.
Journal of the American Society of Nephrology : JASN, 28(1), 166-184 (2016-06-12)
Podocyte injury is the inciting event in primary glomerulopathies, such as minimal change disease and primary FSGS, and glucocorticoids remain the initial and often, the primary treatment of choice for these glomerulopathies. Because inflammation is not readily apparent in these
Hang Mei et al.
International journal of biological sciences, 20(9), 3317-3333 (2024-07-12)
The glomerular podocyte, a terminally differentiated cell, is crucial for the integrity of the glomerular filtration barrier. The re-entry of podocytes into the mitotic phase results in injuries or death, known as mitotic catastrophe (MC), which significantly contributes to the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service