Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500391

Sigma-Aldrich

Anti-ZC3H12A antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MCP-1-induced protein, Anti-MCPIP, Anti-MCPIP1, Anti-Zinc finger CCCH-type containing 12A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€409.00

€409.00


Check Cart for Availability


Select a Size

Change View
100 μG
€409.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€409.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 66 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZC3H12A(80149)

Immunogen

ZC3H12A antibody was raised against a 17 amino acid peptide near the center of human ZC3H12A .

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500391.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elżbieta Boratyn et al.
Journal of cellular biochemistry, 117(3), 694-707 (2015-08-27)
The recently discovered MCPIP1 (monocyte chemoattractant protein-induced protein 1), a multidomain protein encoded by the MCPIP1 (ZC3H12A) gene, has been described as a new differentiation factor, a ribonuclease, and a deubiquitination-supporting factor. However, its role in cancer is poorly recognized.
Zhixing Wang et al.
Cancers, 12(4) (2020-04-11)
RNA-binding proteins (RBPs) play important roles in many cancer types. However, RBPs have not been thoroughly and systematically studied in gliomas. Global analysis of the functional impact of RBPs will provide a better understanding of gliomagenesis and new insights into
Anna Skalniak et al.
Oncology reports, 31(5), 2385-2392 (2014-03-15)
The importance of monocyte chemotactic protein-1-induced protein 1 (MCPIP1) in the negative regulation of inflammatory reactions has already been extensively studied. However, its role in cancer is not yet established. We studied MCPIP1 gene expression in primary human neuroblastomas and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service