Skip to Content
Merck
All Photos(2)

Key Documents

23-025-M

Sigma-Aldrich

UbcH4 Protein, active, 50 µg

Active, full length, recombinant human UbcH4. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.

Synonym(s):

Ubiquitin-conjugating enzyme E2 D2, Ubiquitin-protein ligase D2, UbcH5b, Ubiquitin-conjugating enzyme E2-17kDa 2, UBE2D2

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€207.00

€207.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
50 μG
€207.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€207.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

human

Quality Level

recombinant

expressed in E. coli

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable

NCBI accession no.

General description

C-terminal cMyc and 6His-tagged, recombinant human Smac/DIABLO amino acids 56-end, expressed in E.coli
Ubiquitin-conjugating (E2) enzymes are characterized by the presence of a highly conserved ubiquitin-conjugating domain which accommodates ATP-activated ubiquitin (Ub) via a covalently linked thioester onto its active-site residue. E2 enzymes act via selective protein-protein interactions with the ubiquitin-activating E1 enzyme and ubiquitin ligase E3 enzymes and are able to differentiate effects on downstream substrates, either with a single Ub molecule or a Ub chain. While E3s are involved in substrate selection, E2s are the main determinants for selection of the lysine to construct Ub chains, which thereby directly control the cellular fate of the substrate. The UbcH5 family (UbcH5a, UbcH5b/UbcH4 and UbcH5c) have been shown to be the most active class of E2 enzymes in cell extracts and are associated with the regulation of a number of proteins associated with cell signalling, including p53, IκBα and β-catenin.

Application

Active, full length, recombinant human UbcH4. N-terminal HA, 6His-tagged. For use in Enzyme Assays. Functions as an E2 enzyme in ubiquitination assays.
Ubiquitination Cascade Comoponent: E2

Physical form

Purified using immobilized metal affinity chromatography.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service