Skip to Content
Merck
All Photos(5)

Key Documents

SAB2702380

Sigma-Aldrich

Monoclonal Anti-HEC1 antibody produced in mouse

clone 9G3.23, affinity isolated antibody

Synonym(s):

HEC, HEC1, HsHec1, KNTC2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€401.00

€401.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€401.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€401.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

9G3.23, monoclonal

form

buffered aqueous solution

mol wt

80kDa

species reactivity

hamster, kangaroo rat, human

concentration

1mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
proximity ligation assay: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDC80(10403)

Related Categories

Immunogen

Human HEC1 protein consisting of amino acids 56-642.

Application

Suggested starting dilutions are as follows: FACS: Assay-dependent dilution, ICC/IF: 1:100-1:1000, IHC: Assay-dependent dilution, IP: Assay-dependent dilution, WB: 1:500-1:3000, PLA: Assay-dependent dilution. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Human HEC 1 is a coiled-coil protein that has been demonstrated to have a role in M-phase progression. It is essential for proper chromosome segregation. Its depletion results inthe impairment of chromosome congression and results in spindle checkpoint activation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Other Notes

Purification: Protein G Affinity Purified

Physical form

Phosphate-buffered saline, no preservative added.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Geethu E Thomas et al.
Nature communications, 7, 11665-11665 (2016-05-27)
Kinetochore couples chromosome movement to dynamic microtubules, a process that is fundamental to mitosis in all eukaryotes but poorly understood. In vertebrates, spindle-kinetochore-associated (Ska1-3) protein complex plays an important role in this process. However, the proteins that stabilize Ska-mediated kinetochore-microtubule

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service