Skip to Content
Merck
All Photos(5)

Key Documents

SAB2701957

Sigma-Aldrich

Anti-PIK3CA antibody produced in rabbit

Synonym(s):

MCM, MCMTC, PI3K, p110-alpha

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€471.00

€471.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€471.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€471.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

liquid

mol wt

124 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 500-3000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PIK3CA(5290)

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 3 and 170 (P42336) of PI3 kinase p110 alpha

Application

Suggested starting dilutions are as follows: ICC/IF: 1:100-1:1000, IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Biochem/physiol Actions

Phosphatidylinositol 3-kinase is composed of an 85 kDa regulatory subunit and a 110 kDa catalytic subunit. The protein encoded by this gene represents the catalytic subunit, which uses ATP to phosphorylate PtdIns, PtdIns4P and PtdIns(4,5)P2. This gene has been found to be oncogenic and has been implicated in cervical cancers. [provided by RefSeq]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

1XPBS, 20% Glycerol (pH7). 0.025% ProClin 300 was added as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xian-Fu Ke et al.
Biochemical and biophysical research communications, 450(4), 1297-1303 (2014-07-06)
The pathogenesis of endotoxin-induced acute lung injury (ALI) remains obscure and has not been well elucidated hitherto. Recently, microRNAs have distinct expression profiles in innate immunity, inflammation, and infection. However, the functions of microRNAs in ALI remain unknown. In this
Chia-Lung Chuang et al.
Human molecular genetics, 23(21), 5649-5658 (2014-06-12)
Leucine-rich repeat kinase 2 (LRRK2) is a complex kinase and mutations in LRRK2 are perhaps the most common genetic cause of Parkinson's disease (PD). However, the identification of the normal physiological function of LRRK2 remains elusive. Here, we show that
Bo Liu et al.
PloS one, 9(4), e93576-e93576 (2014-04-15)
Endothelin-1 (ET-1)/endothelin A receptor (ETAR) signaling is important for osteosarcoma (OS) progression. Monoclonal antibodies (mAbs) targeting ganglioside GD2 reportedly inhibit tumor cell viability independent of the immune system. A recent study suggests that ganglioside GD2 may play an important role
Pei-Chien Tsai et al.
Cell biochemistry and function, 32(6), 485-495 (2014-06-27)
The hepatocyte growth factor (HGF)/c-Met signalling pathway is deregulated in most cancers and associated with a poor prognosis in breast cancer. Cardiotoxin III (CTX III), a basic polypeptide isolated from Naja naja atra venom, has been shown to exhibit anticancer
Xiehua Xue et al.
Neuroscience letters, 558, 14-19 (2013-10-26)
We evaluated the neuroprotective effect of electro-acupuncture (EA) on cerebral ischemia-reperfusion (IR) injury and deeply investigated the relationship between this neuroprotective effect and PI3K/Akt pathway. Rats underwent focal cerebral IR injured by suture method and received the in vivo therapeutic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service