Skip to Content
Merck
All Photos(2)

Key Documents

SAB2103980

Sigma-Aldrich

Anti-MPST antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-MGC24539, Anti-MST, Anti-TST2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€423.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

33 kDa

species reactivity

pig, dog, human, rabbit, goat, bovine, horse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MPST(4357)

Immunogen

Synthetic peptide directed towards the middle region of human MPST

Biochem/physiol Actions

MPST transfer of a sulfur ion to cyanide or to other thiol compounds. MPST also has weak rhodanese activity. MPST may have a role in cyanide degradation or in thiosulfate biosynthesis.This gene encodes a protein which can function as a monomer or as a disulfide-linked homodimer and which catalyzes the transfer of a sulfur ion from 3-mercaptopyruvate to cyanide or other thiol compounds. It may be involved in cyanide degradation and in thiosulfate biosynthesis. The encoded cytoplasmic protein is a member of the rhodanese family but is not rhodanese itself, which is a mitochondrial protein. Alternatively spliced transcript variants encoding the same protein have been identified.

Sequence

Synthetic peptide located within the following region: DPAFIKTYEDIKENLESRRFQVVDSRATGRFRGTEPEPRDGIEPGHIPGT

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lamei Yu et al.
Peptides, 111, 89-97 (2018-04-24)
Hydrogen sulfide (H2S) is normally produced from l-cysteine in mammalian tissues and related to the pathogenesis of cardiovascular diseases. The aim of this study is to investigate the effects of H2S donor on atrial natriuretic peptide (ANP) secretion and define

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service