Skip to Content
Merck
All Photos(3)

Documents

MABS1171

Sigma-Aldrich

Anti-MPST Antibody, clone 1H10.1

clone 1H10.1, from mouse

Synonym(s):

3-mercaptopyruvate sulfurtransferase, MpST, MST, tRNA thiouridine modification protein, TUM1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H10.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MPST(4357)

General description

3-mercaptopyruvate sulfurtransferase (EC 2.8.1.2; UniProt P25325; also known as MpST, MST, tRNA thiouridine modification protein, TUM1) is encoded by the MPST (also known as TST2) gene (Gene ID 4357) in human. 3-mercaptopyruvate sulfurtransferase (MpST) catalyzes the transfer of a sulfur ion to cyanide or other thiol compounds and plays a central role in both cysteine degradation and cyanide detoxification. MpST is also known as tRNA thiouridine modification protein (TUM1) and is implicated in other cellular processes in addition to cyanide detoxification, ranging from thiolation of cytosolic tRNAs to the generation of H2S as signaling molecule both in mitochondria and the cytosol. Chronic cyanide intoxications caused by cyanogenic substances present in food products can lead to neurological disorders. MpST and rhodanese [EC 2.8.1.1] are two enzymes that catalyze cyanide detoxification, with MpST also displaying weak rhodanese activity. Cyanide detoxification by rhodanese requires the presence of compounds bearing labile sulfane sulfur, while mercaptopyruvate is needed for MpST catalyzed reaction. Under oxidative stress conditions, the catalytic cysteine site is converted to a sulfenate, resulting in an inactivation of MpST enzymatic activity. Reduced thioredoxin reactivates the enzyme by cleaving an intersubunit disulfide bond to turn the redox switch on. Aberrant MpST activity is found in a few cases of mercaptolactate-cysteine disulfiduria (MCDU) characterized by the appearance of large quantaties of the sulfur-containing amino acid, beta-mercaptolactate-cysteine disulfide, in the urine.

Specificity

Clone 1H10.1 targets an epitope in the second Rhodanese domain present in both spliced isoforms of human MPST reported by UniProt (P25325).

Immunogen

Epitope: Second Rhodanese domain.
GST-tagged recombinant fragment from the second Rhodanese domain of human MPST.

Application

Research Category
Signaling
Research Sub Category
Toxicology & Drug Resistance
This Anti-MPST Antibody, clone 1H10.1 is validated for use in Western Blotting, Immunohistochemistry (Paraffin) for the detection of MPST.
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected MPST in 10 µg of human liver tissue lysate.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected MPST in human heart and kidney tissue sections.

Quality

Evaluated by Western Blotting in SHSY-5Y cell lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected MPST in 10 µg of SHSY-5Y cell lysate.

Target description

~33 kDa observed. 33.05 kDa (isoform 1) and 35.12 kDa (isoform 2) calculated (Met1 removed). Uncharacterized band(s) may appear in some lysates.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anindya Dey et al.
Redox biology, 68, 102958-102958 (2023-11-11)
Astrocytic dysfunction is central to age-related neurodegenerative diseases. However, the mechanisms leading to astrocytic dysfunction are not well understood. We identify that among the diverse cellular constituents of the brain, murine and human astrocytes are enriched in the expression of
Anindya Dey et al.
Science advances, 6(27) (2020-09-17)
The stringent expression of the hypoxia inducible factor-1α (HIF-1α) is critical to a variety of pathophysiological conditions. We reveal that, in normoxia, enzymatic action of cystathionine β-synthase (CBS) produces H2S, which persulfidates prolyl hydroxylase 2 (PHD2) at residues Cys21 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service