Skip to Content
Merck
All Photos(4)

Key Documents

SAB1402736

Sigma-Aldrich

Monoclonal Anti-CRABP2 antibody produced in mouse

clone 4F2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CRABP-II, RBP6

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€418.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€418.00


Ships within 2 weeks. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4F2, monoclonal

form

buffered aqueous solution

mol wt

antigen ~42.7 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRABP2(1382)

General description

A number of specific carrier proteins for members of the vitamin A family have been discovered. Cellular retinoic acid binding proteins (CRABP) are low molecular weight proteins whose precise function remains unknown. The inducibility of the CRABP2 gene suggests that this isoform is important in retinoic acid-mediated regulation of human skin growth and differentiation. It has been postulated that the CRABP2 gene is transcriptionally regulated by a newly synthesized regulatory protein. (provided by RefSeq)

Immunogen

CRABP2 (NP_001869.1, 1 a.a. ~ 138 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPNFSGNWKIIRSENFEELLKVLGVNVMLRKIAVAAASKPAVEIKQEGDTFYIKTSTTVRTTEINFKVGEEFEEQTVDGRPCKSLVKWESENKMVCEQKLLKGEGPKTSWTRELTNDGELILTMTADDVVCTRVYVRE

Application

Monoclonal Anti-CRABP2 antibody produced in mouse is suitable for indirect ELISA and western blot applications.

Biochem/physiol Actions

Cellular retinoic acid binding protein 2 (CRABP2) plays a crucial role in acceleration of the transcriptional activity. In the nucleus, by binding directly to the nuclear receptor RAR, it forms a complex for connecting retinoic acid (RA) from the binding protein to the receptor. This interaction finally helps to accelerate the transcriptional activity. It may have impact in the immature phenotype in glioma cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benito Campos et al.
International journal of cancer, 131(8), 1963-1968 (2012-01-26)
Impairment of endogenous differentiation pathways like retinoic acid (RA) signaling seems to be a central pathogenetic event in astrocytic gliomas. Among others, expression of the differentiation-promoting RA chaperon protein cellular retinoic acid binding protein 2 (CRABP2) is extenuated in high-grade
Avijit Majumdar et al.
The Journal of biological chemistry, 286(49), 42749-42757 (2011-10-15)
Cellular retinoic acid-binding protein II (CRABP-II) undergoes nuclear translocation upon binding of retinoic acid (RA). In the nucleus, CRABP-II directly binds to the nuclear receptor RAR to form a complex through which RA is "channeled" from the binding protein to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service