Skip to Content
Merck
All Photos(2)

Key Documents

HPA017203

Sigma-Aldrich

Anti-CRABP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CRABP-I, Anti-Cellular retinoic acid- binding protein I, Anti-Cellular retinoic acid-binding protein 1, Anti-Retinoic acid-binding protein I, cellular

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QFYIKTSTTVRTTEINFKVGEGFEEETVDGRKCRSLATWENENKIHCTQTLLEGDGPKTYWTRELANDELILTFGADDVVCTRIYVRE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CRABP1(1381)

General description

CRABP1 (cellular retinoic acid-binding protein 1) is a member of the family of fatty acid-binding proteins with high binding affinity for retinoic acid (RA). It is a 15.4kDa protein localized in the cytoplasm.

Immunogen

Cellular retinoic acid-binding protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-CRABP1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

CRABP1 (cellular retinoic acid-binding protein 1) is mainly involved in the regulation of RA (retinoic acid) activity as well as in neurogenesis. It influences the RA-mediated differentiation and proliferation processes. It acts as a tumor suppressor gene in the esophageal carcinogenesis. The characteristics of CRABP1 protein in identifying the serous and clear cell ovarian adenocarcinoma have been reported in several studies.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70170

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yaroslav Kainov et al.
Cell cycle (Georgetown, Tex.), 13(10), 1530-1539 (2014-03-15)
CRABP1 (cellular retinoic acid binding protein 1) belongs to the family of fatty acid binding proteins. Retinoic acid binding is the only known functional activity of this protein. The role of CRABP1 in human carcinogenesis remains poorly understood. Here, for
K Tanaka et al.
Oncogene, 26(44), 6456-6468 (2007-04-18)
Epigenetic alterations and the resulting inactivation of tumor suppressor genes often contribute to the development of various cancers. To identify novel candidates that may be silenced by aberrant methylation in esophageal squamous-cell carcinoma (ESCC), we analysed ESCC cell lines by
Ugljesa Djuric et al.
Molecular & cellular proteomics : MCP, 16(9), 1548-1562 (2017-07-09)
Mass spectrometry (MS) analysis of human post-mortem central nervous system (CNS) tissue and induced pluripotent stem cell (iPSC)-based directed differentiations offer complementary avenues to define protein signatures of neurodevelopment. Methodological improvements of formalin-fixed, paraffin-embedded (FFPE) protein isolation now enable widespread
Irina Favorskaya et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(10), 10295-10300 (2014-07-19)
The impairment of retinoic acid (RA)-dependent signaling is a frequent event during carcinogenesis. Cellular retinoic acid-binding proteins (CRABP1 and CRABP2) are important modulators of RA activity. Up to date, the role of these proteins in cancer progression remains poorly investigated.
Kristine Faye R Pobre et al.
Protein science : a publication of the Protein Society, 28(7), 1324-1339 (2019-05-11)
The effect of mutations in individual proteins on protein homeostasis, or "proteostasis," can in principle depend on the mutations' effects on the thermodynamics or kinetics of folding, or both. Here, we explore this issue using a computational model of in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service