Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306388

Sigma-Aldrich

ANTI-ACTIVIN RECEPTOR TYPE IA (ACVR1) (CENTER R147) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

ACVR1, ACVRLK2, Activin receptor type I, Activin receptor type-1, Activin receptor-like kinase 2

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€457.00

€457.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
400 μL
€457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€457.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

57153 Da

species reactivity

human, mouse

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACVR1(90)

General description

Activin A receptor type 1 (ACVR1) gene encodes a transmembrane protein that belongs to the bone morphogenetic protein (BMP) type I receptors subfamily of the transforming growth factor-β (TGF-β) receptors. ACVR1 is a single-pass type I membrane protein with a ligand-binding extracellular domain, a transmembrane domain, and a cytoplasmic domain. ACVR1 gene is expressed abundantly in endothelial cells, parenchymal cells, fibroblasts, and tumor-derived epithelial cells. This gene is located on human chromosome 2q24.1.

Application

Anti-Activin receptor type IA (ACVR1) (CENTER R147) antibody produced in rabbit has been used in immunohistochemistry (1:100) and western blot (1:100).

Biochem/physiol Actions

Activin A receptor type 1 (ACVR1) protein can bind activin in concert with activin receptor type 2 (ActRII) or activin receptor type-2B (ActRIIB). It participates in activin signaling. It is also capable of forming a complex with either the bone morphogenetic protein (BMP)-2/7-bound bone morphogenetic protein receptor type II (BMPR-II) or activin A receptor type 2A/2B (ACVR2A/ACVR2B). Somatic mutations of the ACVR1 gene are associated with primary ovary and colon cancer. It is considered a target gene to fibrodysplasia ossificans progressiva (FOP).

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ACVR1 (activin A receptor, type I)
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2013)
Diana Rigueur et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 30(4), 733-741 (2014-11-22)
Bone morphogenetic proteins (BMPs) are crucial regulators of chondrogenesis. BMPs transduce their signals through three type I receptors: BMPR1A, BMPR1B, and ACVR1/ALK2. Fibrodysplasia ossificans progressiva (FOP), a rare disorder characterized by progressive ossification of connective tissue, is caused by an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service