Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | € 295,00 |
5 mg | Check Cart for Availability | € 755,00 |
About This Item
Product Name
Sulprostone, ≥95% (HPLC), oil
Quality Segment
assay
≥95% (HPLC)
form
oil
color
colorless to yellow-brown
solubility
DMSO: >10 mg/mL
shipped in
wet ice
storage temp.
−20°C
SMILES string
CS(=O)(=O)NC(=O)CCC\C=C/C[C@@H]1[C@@H](\C=C\[C@@H](O)COc2ccccc2)[C@H](O)CC1=O
InChI
1S/C23H31NO7S/c1-32(29,30)24-23(28)12-8-3-2-7-11-19-20(22(27)15-21(19)26)14-13-17(25)16-31-18-9-5-4-6-10-18/h2,4-7,9-10,13-14,17,19-20,22,25,27H,3,8,11-12,15-16H2,1H3,(H,24,28)/b7-2-,14-13+/t17-,19-,20-,22-/m1/s1
InChI key
UQZVCDCIMBLVNR-TWYODKAFSA-N
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| description ≥95% (HPLC), oil | description ≥98% (HPLC) | description ≥98% (HPLC), synthetic | description ≥98% (HPLC) |
| form oil | form oil | form (powder to crystals) | form powder |
| assay ≥95% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (HPLC) |
| Quality Level 100 | Quality Level - | Quality Level 200 | Quality Level - |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| solubility DMSO: >10 mg/mL | solubility - | solubility ethanol: 5 mg/mL | solubility H2O: 2 mg/mL, clear |
| color colorless to yellow-brown | color colorless to yellow | color - | color white to beige |
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





