Key Documents
S6690
Sigma-Aldrich
Nunc® Lab-Tek® II - CC2 Chamber Slide system
4 wells, glass slide, 1.7 cm2/well, sterile
Synonym(s):
cell culture chamber, chamber slides, microscope slides, microscopy slides
Select a Size
1 PKG
€471.00
€471.00
Estimated to ship on30 March 2025
Select a Size
Change View
1 PKG
€471.00
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
€471.00
Estimated to ship on30 March 2025
material
colorless
polystyrene chamber
sterility
sterile
sterile
feature
CE marked
manufacturer/tradename
Nunc 154917
packaging
pkg of carton of 16
well area
1.7 cm2
wells
4
working volume
0.5-1.0 mL
suitability
suitable for (cell culture; optimal for culturing fastidious cells (e.g. neurons))
General description
Nunc® Lab-Tek II® - CC2 Chamber Slide System
  • Chemically coated growth surface on glass slide that mimics polylysine
  • Provides binding sites optimal for fastidious cells (e.g. neurons)
  • Growth surface remains stable without refrigeration
  • Light blue frosted writing area
  • Slide seperator included in each pack
  • CE marked
Legal Information
CC2 is a trademark of Thermo Fisher Scientific or its subsidiaries
Chamber Slide is a trademark of Thermo Fisher Scientific or its subsidiaries
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)