Skip to Content
Merck
All Photos(1)

Key Documents

S6268

Sigma-Aldrich

Sigma Pasteur pipettes

non-plugged, L 9 in.

Synonym(s):

Pasteur Pipette

Sign Into View Organizational & Contract Pricing

Select a Size

250 EA
€84.70
1000 EA
€306.00

€84.70


Estimated to ship on11 April 2025



Select a Size

Change View
250 EA
€84.70
1000 EA
€306.00

About This Item

UNSPSC Code:
41121500
eCl@ss:
32079210
NACRES:
NB.01

€84.70


Estimated to ship on11 April 2025


material

glass

sterility

sterile; γ-irradiated
sterile; γ-irradiated

feature

graduations: no

packaging

pack of 25 ea (packed in blisterpacks)

manufacturer/tradename

Sigma-Aldrich

L

9 in.

Looking for similar products? Visit Product Comparison Guide

Application

Sigma Pasteur pipettes have been used to transfer embryos and manipulate human embryonic stem cell lines (hESC).[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derivation of Human Embryonic Stem Cell Lines from Poor Quality Embryos
Ronak S, et al.
Methods in Molecular Biology, 151?161-151?161 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service