Skip to Content
Merck
All Photos(2)

Documents

S4257

Sigma-Aldrich

Sphingosylphosphorylcholine

≥98%, powder

Synonym(s):

(2S,3R,4E)-2-Amino-4-octadecene-3-hydroxy-1-phosphocholine, 2-[[[(2-Amino-3-hydroxy-4-octadecenyl)oxy]hydroxy-phosphinyl]oxy]-N,N,N-trimethylethanaminium inner salt, D-erythro-Sphingosine phosphocholine, Lyso SM (d18:1), Lysosphingomyelin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H49N2O5P
CAS Number:
Molecular Weight:
464.62
Beilstein:
7342520
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98%

form

powder

color

light yellow

solubility

chloroform/methanol (2:1): complete 10 mg/ml, clear to slightly hazy, colorless to light yellow

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(N)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C23H49N2O5P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23(26)22(24)21-30-31(27,28)29-20-19-25(2,3)4/h17-18,22-23,26H,5-16,19-21,24H2,1-4H3/b18-17+/t22-,23+/m0/s1

InChI key

JLVSPVFPBBFMBE-HXSWCURESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Neurotrophic effects of Sphingosylphosphorylcholine were studied in cerebellar granule neurons of mice and PC-12 cells.

Biochem/physiol Actions

Sphingosylphosphorylcholine (SPC) is a bioactive lipid that mediates intracellular and extracellular signaling. It mobilizes Ca2+ from intracellular stores via an IP3-independent pathway. It stimulates the production of CCL2 that may contribute to development of atherosclerosis. SPC is a ligand for endothelial differentiation gene receptor 3 (EDG3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Uehara et al.
FEBS letters, 460(3), 467-471 (1999-11-11)
The effects of the lysosphingolipid, sphingosylphosphorylcholine (SPC), on the cardiac ryanodine receptor (RyR) were examined. The open probability of cardiac RyR incorporated in lipid bilayers was decreased by cytoplasmic, but not lumenal side application of micromolar concentrations of SPC. Modification
Naoko Konno et al.
Biochemical and biophysical research communications, 364(1), 163-168 (2007-10-16)
Sphingosylphosphorylcholine (SPC) is a choline-containing naturally occurring derivative of sphingolipid involved in various biological processes. Here we show that SPC displays neurotrophic effects in cerebellar granule neurons (CGNs) and in PC12 cells. When CGNs were cultured under non-depolarizing condition, they
H Okamoto et al.
Biochemical and biophysical research communications, 260(1), 203-208 (1999-06-25)
AGR16/H218/EDG5 and EDG1 are functional receptors for lysosphingolipids, whereas EDG2 and EGD4 are receptors for lysophosphatidic acid (LPA). The present study demonstrates that EDG3, the yet poorly defined member of the EDG family G protein-coupled receptors, shows identical agonist specificity
D Meyer zu Heringdorf et al.
FEBS letters, 410(1), 34-38 (1997-06-23)
Sphingolipid breakdown products are now being recognized to play a dual role in cellular signalling, acting as intracellular as well as extracellular signalling molecules. Both types of action may even be found with one sphingolipid species. The recent demonstration of
Ha Young Lee et al.
Journal of immunology (Baltimore, Md. : 1950), 186(7), 4347-4353 (2011-03-04)
Sphingosylphosphorylcholine (SPC) is a component of high-density lipoprotein particles. We investigated the functional role of SPC in HUVECs. SPC stimulation induced production of the CCL2 chemokine in a PTX-sensitive G-protein-dependent manner. SPC treatment caused the activation of NF-κB and AP-1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service