Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mL | Check Cart for Availability | € 179,00 |
About This Item
conjugate
phycoerythrin (R-PE) conjugate
Quality Segment
form
buffered aqueous solution
mol wt
~60 kDa (four identical subunits)
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5 μg/mL using human appendix or human tonsil tissue
shipped in
wet ice
storage temp.
2-8°C
General description
Application
Physical form
Preparation Note
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) immunohistochemistry (formalin-fixed, paraffin-embedded sections): 5 μg/mL using human appendix or human tonsil tissue | technique(s) direct ELISA: 1:50,000 | technique(s) - | technique(s) - |
| form buffered aqueous solution | form lyophilized powder | form lyophilized powder | form liquid |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. −20°C | storage temp. −20°C | storage temp. 2-8°C |
| shipped in wet ice | shipped in - | shipped in - | shipped in ambient |
| conjugate phycoerythrin (R-PE) conjugate | conjugate peroxidase conjugate | conjugate alkaline phosphatase conjugate | conjugate - |
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



