Skip to Content
Merck
All Photos(1)

Key Documents

189737

Sigma-Aldrich

Streptavidin, Phycoerythrin Conjugate

This Streptavidin, Phycoerythrin Conjugate is validated for use in Enzyme Immunoassay, Flow Cytometry.

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€390.00

€390.00


Estimated to ship on20 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€390.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

€390.00


Estimated to ship on20 May 2025


Request a Bulk Order

Quality Level

form

liquid

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

do not freeze
protect from light

fluorescence

λex 500 nm
λem 575 nm

shipped in

ambient

storage temp.

2-8°C

General description

Streptavidin conjugated to R-phycoerythrin. Specific for biotin, biotinylated proteins, or biotinylated antibodies.

Application

Enzyme Immunoassay (see comments)

Flow Cytometry (1-3 µg per 1 x 10⁶ cells in 100 µl)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 150 mM NaCl, 50 mM sodium phosphate, 1% BSA, 0.1% glycerol, pH 7.6. Ammonium sulfate free.

Reconstitution

Do not dilute the entire vial at one time. Dilute only as needed in 50 mM sodium phosphate buffer, pH 7.2, 2-5 mg/ml of BSA or other physiological buffer.

Other Notes

Lindmo, T., et al. 1990. J. Immunol. Methods126, 183.
Starling, J.J., et al. 1988. Cancer Res.48, 6211.
The phycobiliprotein used in making this conjugate had a M.W. of 240,000. Molar extinction coefficient at 565 nm: 1.9 x 106. Excitation Max: 500 nm emission max: 575 nm.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service