Skip to Content
Merck
All Photos(4)

Key Documents

RDD014

Sigma-Aldrich

BIS-TRIS propane

anhydrous, free-flowing, Redi-Dri, ≥99.0%

Synonym(s):

1,3-Bis[tris(hydroxymethyl)methylamino]propane

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€249.00
500 G
€794.00
1 KG
€1,410.00

€249.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€249.00
500 G
€794.00
1 KG
€1,410.00

About This Item

Linear Formula:
CH2[CH2NHC(CH2OH)3]2
CAS Number:
Molecular Weight:
282.33
Beilstein:
1786109
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

€249.00


Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99.0%

form

powder

quality

free-flowing

impurities

≤0.2% water (Karl Fischer)
≤0.5% tris(hydroxymethyl aminomethane (H-NMR)

pH

9.0-10.1 (20-25 °C, 1 M in water)

useful pH range

6.3-9.5

pKa (25 °C)

(1) 6.8, (2) 9.0

mp

164-165 °C (lit.)

solubility

H2O: 30 % (w/w), clear, colorless

cation traces

heavy metals (as Pb): ≤5 ppm

absorption

≤0.05 at 290 at 30 % (w/w)

SMILES string

OCC(CO)(CO)NCCCNC(CO)(CO)CO

InChI

1S/C11H26N2O6/c14-4-10(5-15,6-16)12-2-1-3-13-11(7-17,8-18)9-19/h12-19H,1-9H2

InChI key

HHKZCCWKTZRCCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Comparing solution-gate and bottom-gate nanowire field-effect transistors on pH sensing with different salt concentrations and surface modifications.: This study investigates the pH sensing performance of nanowire field-effect transistors with varying salt concentrations and surface modifications, using BIS-TRIS propane as a buffer. The findings highlight the importance of buffer choice in optimizing sensor sensitivity and stability (Hu et al., 2024).

  • Laser-induced tuning of graphene field-effect transistors for pH sensing.: This research explores the use of BIS-TRIS propane in the laser-induced tuning of graphene field-effect transistors for enhanced pH sensing capabilities. The study demonstrates improved sensor accuracy and response times (Lampinen et al., 2023).

  • Protein association on multimodal chromatography media.: This study utilizes BIS-TRIS propane in multimodal chromatography to investigate protein associations, aiming to enhance the efficiency of protein purification processes (Muca and Antos, 2023).

  • Novel anion exchange membrane chromatography method for the separation of empty and full adeno-associated virus.: BIS-TRIS propane is employed in a novel anion exchange membrane chromatography method to separate empty and full adeno-associated viruses, demonstrating its effectiveness in biotechnological applications (Hejmowski et al., 2022).

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service