Skip to Content
Merck

Skip To

RAB0580

Canine VEGFA / Vascular Endothelial Growth Factor A ELISA Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
96 wells
Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
€ 878,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
canine ... VEGFA(403802)
Input:
sample type plasma
sample type serum
sample type cell culture supernatant(s)
Species reactivity:
canine
Storage temp.:
−20°C
Shipped in:
wet ice

€ 878,00


Usually Ships in 4 Weeks. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

canine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

canine ... VEGFA(403802)

General description

The antibody pair provided in this kit recognizes Canine VEGF.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0859RAB0318RAB0821
description

-

description

-

description

for serum, plasma and cell culture supernatants

description

-

Gene Information

canine ... VEGFA(403802)

Gene Information

mouse ... Flt4(14257)

Gene Information

canine ... IL8(403850)

Gene Information

mouse ... Ifnz(319146)

species reactivity

canine

species reactivity

mouse

species reactivity

canine

species reactivity

mouse

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.2 ng/mL
standard curve range: 0.205-50 ng/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6.2 pg/mL
standard curve range: 6.14-1500 pg/mL

input

sample type plasma
sample type serum
sample type cell culture supernatant(s)

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

capture ELISA: suitable

technique(s)

ELISA: suitable

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters