Skip to Content
Merck

Skip To

RAB0318

Canine CXCL8 / IL-8 ELISA Kit

for serum, plasma and cell culture supernatants

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 533,00

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Gene information:
canine ... IL8(403850)
Input:
sample type cell culture supernatant(s)
sample type serum
sample type plasma
Species reactivity:
canine
Storage temp.:
−20°C
Shipped in:
wet ice

€ 533,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

canine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

canine ... IL8(403850)

General description

The Canine IL-8 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of canine IL-8 in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Canine IL-8

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0321RAB0301RAB0295
species reactivity

canine

species reactivity

porcine

species reactivity

rat

species reactivity

mouse

Gene Information

canine ... IL8(403850)

Gene Information

Porcine ... IL8(396880)

Gene Information

rat ... Il4(287287)

Gene Information

mouse ... Il3(16187)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 6 pg/mL
standard curve range: 6.14-1500 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 10 pg/mL
standard curve range: 8.19-2000 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.5 pg/mL
standard curve range: 0.66-160 pg/mL

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.2 pg/mL
standard curve range: 0.16-40 pg/mL

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type cell culture supernatant(s)
sample type serum
sample type plasma

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric

detection method

colorimetric


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • ELISA 5X Assay/Sample Diluent Buffer E (Item E2)
    SDS

  • ELISA Stop Solution (Item I)
    SDS

  • ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)
    SDS

  • 20X Wash Buffer (Item B)
    SDS

pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters