Skip to Content
Merck
All Photos(1)

Documents

RAB0550

Sigma-Aldrich

Human TNFRSF11A ELISA Kit

for serum, plasma, cell culture supernatants and urine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type urine
sample type cell culture supernatant(s)
sample type serum
sample type plasma

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.4 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

General description

The antibody pair provided in this kit detects Human TNFRSF11A in serum, plasma, cell culture supernatants, and urine.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keisha M Cawley et al.
Cell reports, 32(10), 108052-108052 (2020-09-10)
Osteoprotegerin (OPG) inhibits the ability of receptor activator of nuclear factor κB (NF-κB) ligand (RANKL) to stimulate the differentiation, activity, and survival of bone-resorbing osteoclasts. Genetic studies in mice show that osteocytes are an important source of RANKL, but the
Yuka Yashima et al.
Biomedical research (Tokyo, Japan), 41(2), 91-99 (2020-04-21)
The present study aimed to investigate the effect of a compressive force (CF) on the expression of receptor activator of nuclear factor kappa-B ligand (RANKL), osteoprotegerin (OPG), and vascular endothelial growth factor (VEGF) in murine osteocytes (MLO-Y4) as well as
Abolghasem Hadinia et al.
Clinical laboratory, 66(5) (2020-05-12)
The pathobiology of initiation and progression of nonalcoholic fatty liver disease (NAFLD) has not been completely elucidated. It seems that the RANK/RANKL/OPG cytokine system play an etiologic role in pathogenesis of this disease. This study aimed to investigate the plasma
H Herschel Conaway et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(12), 14394-14409 (2019-11-02)
Glucocorticoid (GC) therapy decreases bone mass and increases the risk of fractures. We investigated interactions between the GC dexamethasone (DEX) and the bone resorptive agents 1,25(OH)2-vitamin D3 (D3) and parathyroid hormone (PTH) on osteoclastogenesis. We observed a synergistic potentiation of
N E Kushlinskii et al.
Bulletin of experimental biology and medicine, 163(4), 478-481 (2017-08-31)
The content of components of the RANK/RANKL/OPG system, the key regulator of homeostasis in the bone tissue, in blood serum samples from 199 patients with primary bone neoplasms and 131 practically healthy volunteers was measured by ELISA. Borderline giantcell tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service