Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery) | € 549,00 |
About This Item
sample type plasma
sample type serum
€ 549,00
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable, capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type plasma
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 3 pg/mL
standard curve range: 1.37-1000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Timp1(21857)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
Please type the word sample in the text box provided for lot number.
1 of 1
This Item | |||
|---|---|---|---|
| description for serum, plasma and cell culture supernatant | description for serum, plasma and cell culture supernatant | description for serum, plasma, cell culture supernatant and urine | description for serum, plasma and cell culture supernatant |
| species reactivity mouse | species reactivity rat | species reactivity human | species reactivity mouse |
| Gene Information mouse ... Timp1(21857) | Gene Information rat ... Timp1(116510) | Gene Information human ... TIMP1(7076) | Gene Information mouse ... Timp2(21858) |
| input sample type cell culture supernatant(s) | input sample type plasma | input sample type cell culture supernatant(s) | input sample type cell culture supernatant(s) |
| assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% | assay range inter-assay cv: <12% |
| technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) capture ELISA: suitable | technique(s) capture ELISA: suitable | technique(s) capture ELISA: suitable |
| detection method colorimetric | detection method colorimetric | detection method colorimetric | detection method colorimetric |
Still not finding the right product?
Explore all of our products under Mouse TIMP-1 ELISA Kit
Kit Components Also Available Separately
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




