Skip to Content
Merck
All Photos(2)

Key Documents

RAB0466

Sigma-Aldrich

Human TIMP-1 ELISA Kit

for serum, plasma, cell culture supernatant and urine

Synonym(s):

Metalloproteinase inhibitor 1, TIMP-1

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€528.00

€528.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
1 KIT
€528.00

About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

€528.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type cell culture supernatant(s)
sample type urine
sample type serum
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 40 pg/mL
standard curve range: 24.69-18000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TIMP1(7076)

Immunogen

Recombinant Human TIMP1

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADAELISA 1X Assay/Sample Diluent Buffer A (Item D1)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chunyan Meng et al.
PloS one, 13(11), e0207039-e0207039 (2018-11-21)
Tissue inhibitor of metalloproteinase-1 (TIMP-1) is a glycoprotein involved in cell survival and tumorigenesis. There have been some promising results regarding the diagnostic value of TIMP-1 for patients with colorectal cancer (CRC). The aim of the present study was to
Xin Zheng et al.
Cellular signalling, 28(9), 1314-1324 (2016-06-15)
Aberrant tumor microenvironment is involved closely in tumor initiation and progression, in which cancer associated fibroblasts (CAFs) play a pivotal role. Both IL-6/STAT3 signaling and TIMP-1 have been found to modulate the crosstalk between tumor cells and CAFs in tumor
Xin Zheng et al.
Cellular signalling, 28(9), 1314-1324 (2016-06-15)
Aberrant tumor microenvironment is involved closely in tumor initiation and progression, in which cancer associated fibroblasts (CAFs) play a pivotal role. Both IL-6/STAT3 signaling and TIMP-1 have been found to modulate the crosstalk between tumor cells and CAFs in tumor
Hirokuni Akahori et al.
Journal of hypertension, 32(7), 1534-1541 (2014-04-25)
Clinical studies have suggested the beneficial effects of statin therapy on diastolic heart failure. However, the mechanism of the beneficial effects of statin on diastolic heart failure remains unknown. We examined the effect of atorvastatin on the cardiac function of
Nejat Nizam et al.
Inflammation, 37(5), 1771-1778 (2014-05-07)
Salivary, serum matrix metalloproteinase-8 (MMP-8), tissue inhibitor of matrix metalloproteinases-1 (TIMP-1), neutrophil elastase (NE), and myeloperoxidase (MPO) levels were investigated in generalized chronic periodontitis (GCP), generalized aggressive periodontitis (GAgP), and healthy groups. Whole-mouth clinical periodontal measurements were recorded. Salivary, serum

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service