Skip to Content
Merck

Skip To

RAB0164

Phospho-EGFR (pTyr1045) and pan-EGFR ELISA Kit

for detection of human phospho-egfr (pTyr1045) and pan-egfr in cell and tissue lysates

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 kit
Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)
€ 715,00

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... EGFR(1965)
Input:
sample type: human tissue lysate
sample type: human cell lysate
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice

€ 715,00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

species reactivity

human

technique(s)

ELISA: suitable, capture ELISA: suitable

input

sample type: human tissue lysate
sample type: human cell lysate

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... EGFR(1965)

General description

The Phospho-EGFR (pTyr1045) ELISA (Enzyme-Linked Immunosorbent Assay) kit is a very rapid, convenient and sensitive assay kit that can monitor the activation or function of important biological pathways in cell lysates.

Immunogen

EGFR (pTyr1045) synthetic phosphopeptide, Recombinant Human EGFR

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
RAB0163RAB0167RAB0170
description

for detection of human phospho-egfr (pTyr1045) and pan-egfr in cell and tissue lysates

description

for detection of human phospho-egfr (pTyr1045) in cell and tissue lysates

description

for detection of human phospho-egfr (pTyr1086) in cell and tissue lysates

description

for detection of human phospho-egfr (pTyr992) and pan-egfr in cell and tissue lysates

Gene Information

human ... EGFR(1965)

Gene Information

human ... EGFR(1965)

Gene Information

human ... EGFR(1965)

Gene Information

human ... EGFR(1965)

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

human

input

sample type: human tissue lysate
sample type: human cell lysate

input

sample type: human cell lysate
sample type: human tissue lysate

input

sample type: human cell lysate
sample type: human tissue lysate

input

sample type: human cell lysate
sample type: human tissue lysate

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

technique(s)

capture ELISA: suitable

technique(s)

ELISA: suitable, capture ELISA: suitable

technique(s)

capture ELISA: suitable

technique(s)

capture ELISA: suitable

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


signalword

Warning

flash_point_f

Not applicable

flash_point_c

Not applicable

pictograms

Corrosion

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters