Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 kit | Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery) | € 664,00 |
About This Item
sample type: human tissue lysate
€ 664,00
Quality Segment
species reactivity
human
technique(s)
ELISA: suitable, capture ELISA: suitable
input
sample type: human cell lysate
sample type: human tissue lysate
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... EGFR(1965)
General description
Immunogen
Application
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
1 of 1
This Item | |||
|---|---|---|---|
| description for detection of human phospho-egfr (pTyr1045) in cell and tissue lysates | description for detection of human phospho-egfr (pTyr1045) and pan-egfr in cell and tissue lysates | description for detection of human phospho-egfr (pTyr1086) in cell and tissue lysates | description for detection of human phospho-egfr (pTyr992) and pan-egfr in cell and tissue lysates |
| Gene Information human ... EGFR(1965) | Gene Information human ... EGFR(1965) | Gene Information human ... EGFR(1965) | Gene Information human ... EGFR(1965) |
| species reactivity human | species reactivity human | species reactivity human | species reactivity human |
| input sample type: human cell lysate | input sample type: human tissue lysate | input sample type: human cell lysate | input sample type: human cell lysate |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable | technique(s) ELISA: suitable, capture ELISA: suitable |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
signalword
Warning
hcodes
pcodes
Hazard Classifications
Met. Corr. 1
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




